Proteintech
Manufacturer Code:150301AP150UL
Catalog # 50-172-9918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CUTC Fusion Protein Ag2126 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI 32 CUTC RP11 483F11.3; Copper homeostasis protein cutC homolog; cutC copper transporter homolog
Gene Aliases: 2310039I18Rik; AI326282; CGI-32; CUTC
UniProt ID: (Human) Q9NTM9, (Mouse) Q9D8X1
Entrez Gene ID: (Human) 51076, (Mouse) 66388, (Rat) 361760
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.