Novus Biologicals
Manufacturer Code:NBP271623AF647
Catalog # N271623A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5B8 |
Immunogen | Full length recombinant protein of human CUGBP1/CELF1 (NP_006551) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 50 kDa nuclear polyadenylated RNA-binding protein; bruno-like 2; Bruno-like protein 2; BRUNOL250 kDa nuclear polyadenylated RNA-binding protein bruno-like 2 Bruno-like protein 2 CUG RNA-binding protein CUG triplet repeat RNA binding protein 1 CUG triplet repeat RNA-binding protein 1 CUG-BP CUG-BP- and ETR-3-like factor 1 CUGBP Elav-like family member 1 CUGBP Elav-like family member 1 CUGBP1CUG triplet repeat RNA-binding protein 1 CUGBPCELF-1 Deadenylation factor CUG-BP EDEN-BP EDEN-BP homolog embryo deadenylation element binding protein Embryo deadenylation element-binding protein homolog hNab50 NAB50CUG-BP1 NAPOR nuclear polyadenylated RNA-binding protein 50-kD RNA-binding protein BRUNOL-2; CELF-1; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA-binding protein 1; CUG-BP- and ETR-3-like factor 1; CUG-BP1; CUGBP Elav-like family member 1; Deadenylation factor CUG-BP; EDEN-BP homolog; embryo deadenylation element binding protein; Embryo deadenylation element-binding protein homolog; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2
Gene Aliases: BRUNOL2; CELF1; CUG-BP; CUGBP; CUGBP1; EDEN-BP; hNab50; NAB50; NAPOR
UniProt ID: (Human) Q92879
Entrez Gene ID: (Human) 10658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.