Invitrogen
Manufacturer Code:MA116675
Catalog # AMPA6675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Gel Shift (GS) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B1 |
Immunogen | CUG-BP1 human nuclear RNA binding protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa nuclear polyadenylated RNA-binding protein; Brain protein F41; bruno-like 2; Bruno-like protein 2; CELF-1; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA-binding protein 1; CUG-BP- and ETR-3-like factor 1; CUG-BP1; CUGBP Elav-like family member 1; CUGBP, Elav-like family member 1-like; Deadenylation factor CUG-BP; Deadenylation factor EDEN-BP; EDEN-BP homolog; embryo deadenylation element binding protein; Embryo deadenylation element-binding protein homolog; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2; RP23-147D3.7 1600010O03Rik AA407467 Brunol2 CUG-BP CUG-BP1 CUGBP D2Wsu101e HNAB50 NAB50
Gene Aliases: 1600010O03Rik; AA407467; BRUNOL2; CELF1; CUG-BP; CUG-BP1; CUGBP; CUGBP1; D2Wsu101e; EDEN-BP; HNAB50; NAB50; NAPOR
UniProt ID: (Human) Q92879, (Mouse) A2AFX0, (Rat) Q4QQT3
Entrez Gene ID: (Human) 10658, (Bovine) 540475, (Pig) 100514512, (Rabbit) 100354668, (Mouse) 13046, (Rat) 362160
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.