Invitrogen
Manufacturer Code:PA536277
Catalog # PIPA536277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 62-115 of human CUG-BP1/2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 50 kDa nuclear polyadenylated RNA-binding protein; Brain protein F41; bruno-like 2; Bruno-like protein 2; Bruno-like protein 3; CELF-1; CELF-2; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat RNA-binding protein 2; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA binding protein 2; CUG triplet repeat, RNA-binding protein 1; CUG-BP- and ETR-3-like factor 1; CUG-BP- and ETR-3-like factor 2; CUG-BP1; CUG-BP2; CUGBP Elav-like family member 1; CUGBP Elav-like family member 1 CELF-1 50 kDa nuclear polyadenylated RNA-binding protein Bruno-like protein 2 CUG triplet repeat RNA-binding protein 1 CUG-BP1 CUG-BP- and ETR-3-like factor 1 Deadenylation factor CUG-BP Embryo deadenylation element-binding protein homolog EDEN-BP homolog RNA-binding protein BRUNOL-2 CELF1 BRUNOL2 CUGBP CUGBP1 NAB50 CUGBP Elav-like family member 2 CELF-2 Bruno-like protein 3 CUG triplet repeat RNA-binding protein 2 CUG-BP2 CUG-BP- and ETR-3-like factor 2 ELAV-type RNA-binding protein 3 ETR-3 Neuroblastoma apoptosis-related RNA-binding protein hNAPOR RNA-binding protein BRUNOL-3 CELF2 BRUNOL3 CUGBP2 ETR3 NAPOR; CUGBP Elav-like family member 2; Cugbp- and Etr3-like factor 2; Deadenylation factor CUG-BP; Deadenylation factor EDEN-BP; EDEN-BP homolog; ELAV-type RNA-binding protein 3; embryo deadenylation element binding protein; Embryo deadenylation element-binding protein homolog; ETR-3; hNAPOR; mNapor; Neuroblastoma apoptosis-related RNA-binding protein; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2; RNA-binding protein BRUNOL-3; RNA-binding protein Brunol3; rNapor
Gene Aliases: 1600010O03Rik; AA407467; B230218O03; B230345P09Rik; BRUNOL2; BRUNOL3; C88023; CELF-2; CELF1; CELF2; CUG-BP; CUG-BP1; CUG-BP2; CUGBP; CUGBP1; CUGBP2; D230046B21Rik; D2Wsu101e; EDEN-BP; ETR-3; ETR3; HNAB50; mETR-3; NAB50; NAPOR; Napor-2
UniProt ID: (Human) Q92879, (Human) O95319, (Mouse) A2AFX0, (Rat) Q4QQT3, (Rat) Q792H5, (Mouse) Q9R0B2
Entrez Gene ID: (Human) 10658, (Human) 10659, (Mouse) 13046, (Rat) 362160, (Rat) 29428, (Mouse) 14007
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.