Invitrogen
Manufacturer Code:PA5115064
Catalog # PIPA5115064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CUGBP2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1600010O03Rik 50 kDa nuclear polyadenylated RNA-binding protein AA407467 B230218O03 B230345P09Rik Brain protein F41 BRUNOL2 BRUNOL3 bruno-like 2 bruno-like protein 2 Bruno-like protein 3 C88023 Celf1 CELF-1 CELF2 CELF-2 CUG RNA-binding protein CUG triplet repeat RNA-binding protein 1 CUG triplet repeat RNA-binding protein 2 CUG triplet repeat RNA binding protein 1 CUG triplet repeat RNA binding protein 2 CUG triplet repeat RNA-binding protein 1 CUGBP CUG-BP CUG-BP- and ETR-3-like factor 1 CUG-BP- and ETR-3-like factor 2 Cugbp- and Etr3-like factor 2 CUGBP Elav-like family member 1 CUGBP Elav-like family member 2 CUGBP Elav-like family member 1 CUGBP Elav-like family member 2 Cugbp1 CUG-BP1 CUGBP2 CUG-BP2 D230046B21Rik D2Wsu101e Deadenylation factor CUG-BP deadenylation factor EDEN-BP EDEN-BP EDEN-BP homolog ELAV-type RNA-binding protein 3 embryo deadenylation element binding protein embryo deadenylation element-binding protein homolog Etr3 ETR-3 hNab50 hNAPOR mETR-3 mNapor NAB50 Napor Napor-2 Neuroblastoma apoptosis-related RNA-binding protein nuclear polyadenylated RNA-binding protein 50-kD RNA-binding protein BRUNOL-2 RNA-binding protein Brunol3 RNA-binding protein BRUNOL-3 rNapor; 50 kDa nuclear polyadenylated RNA-binding protein; Brain protein F41; bruno-like 2; Bruno-like protein 2; Bruno-like protein 3; CELF-1; CELF-2; CUG RNA-binding protein; CUG triplet repeat RNA-binding protein 1; CUG triplet repeat RNA-binding protein 2; CUG triplet repeat, RNA binding protein 1; CUG triplet repeat, RNA binding protein 2; CUG triplet repeat, RNA-binding protein 1; CUG-BP- and ETR-3-like factor 1; CUG-BP- and ETR-3-like factor 2; CUG-BP1; CUG-BP2; CUGBP Elav-like family member 1; CUGBP Elav-like family member 2; Cugbp- and Etr3-like factor 2; Deadenylation factor CUG-BP; Deadenylation factor EDEN-BP; EDEN-BP homolog; ELAV-type RNA-binding protein 3; embryo deadenylation element binding protein; Embryo deadenylation element-binding protein homolog; ETR-3; hNAPOR; mNapor; Neuroblastoma apoptosis-related RNA-binding protein; nuclear polyadenylated RNA-binding protein, 50-kD; RNA-binding protein BRUNOL-2; RNA-binding protein BRUNOL-3; RNA-binding protein Brunol3; rNapor
Gene Aliases: 1600010O03Rik; AA407467; B230218O03; B230345P09Rik; BRUNOL2; BRUNOL3; C88023; CELF-2; CELF1; CELF2; CUG-BP; CUG-BP1; CUG-BP2; CUGBP; CUGBP1; CUGBP2; D230046B21Rik; D2Wsu101e; EDEN-BP; ETR-3; ETR3; HNAB50; mETR-3; NAB50; NAPOR; Napor-2
UniProt ID: (Human) Q92879, (Human) O95319, (Mouse) A2AFX0, (Rat) Q4QQT3, (Rat) Q792H5, (Mouse) Q9R0B2
Entrez Gene ID: (Human) 10658, (Human) 10659, (Mouse) 13046, (Rat) 362160, (Rat) 29428, (Mouse) 14007
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.