Invitrogen
Manufacturer Code:PA520620
Catalog # PIPA520620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide from near the carboxy terminus of human CUEDC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CUE domain-containing protein 2; HOYS6 C10orf66 bA18I14.5 CUE domain-containing protein 2
Gene Aliases: 3010002G01Rik; bA18I14.5; C10orf66; CUEDC2; HOYS6
UniProt ID: (Human) Q9H467, (Mouse) Q9CXX9, (Rat) A1L131
Entrez Gene ID: (Human) 79004, (Mouse) 67116, (Rat) 294009
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.