Novus Biologicals
Manufacturer Code:NBP29209920UL
Catalog # NB026091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 186-515 of human CTU2 (NP_001012777.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C16orf84 chromosome 16 open reading frame 84 cytoplasmic tRNA 2-thiolation protein 2 cytosolic thiouridylase subunit 2 homolog (S. pombe) MGC131681 MGC148063 NCS2MGC148064 UPF0432; Cytoplasmic tRNA 2-thiolation protein 2; Cytosolic thiouridylase subunit 2; cytosolic thiouridylase subunit 2 homolog
Gene Aliases: C16orf84; CTU2; NCS2; UPF0432
UniProt ID: (Human) Q2VPK5
Entrez Gene ID: (Human) 348180
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.