Invitrogen
Manufacturer Code:PA599306
Catalog # PIPA599306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ATPBD3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP binding domain 3; ATP binding domain 3 Atpbd3 ATP-binding domain-containing protein 3 BC005752 Cancer-associated gene protein cancer-associated protein CTU1 Cytoplasmic tRNA 2-thiolation protein 1 cytoplasmic tRNA adenylyltransferase 1 cytosolic thiouridylase subunit 1 cytosolic thiouridylase subunit 1 homolog cytosolic thiouridylase subunit 1 homolog (S. pombe) NCS6; ATP-binding domain-containing protein 3; Cancer-associated gene protein; cancer-associated protein; CTU1; Cytoplasmic tRNA 2-thiolation protein 1; Cytoplasmic tRNA adenylyltransferase 1; cytosolic thiouridylase subunit 1 homolog
Gene Aliases: ATPBD3; CTU1; NCS6
UniProt ID: (Human) Q7Z7A3
Entrez Gene ID: (Human) 90353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.