Abnova
Manufacturer Code:PAB17402
Catalog # 89-119-059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human CTU1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CTU1.
For Research Use Only
Protein Aliases: ATP binding domain 3; ATP-binding domain-containing protein 3; Cancer-associated gene protein; cancer-associated protein; CTU1; Cytoplasmic tRNA 2-thiolation protein 1; Cytoplasmic tRNA adenylyltransferase 1; cytosolic thiouridylase subunit 1 homolog; MGC17332
Gene Aliases: ATPBD3; CTU1; NCS6
UniProt ID: (Human) Q7Z7A3
Entrez Gene ID: (Human) 90353
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.