Invitrogen
Manufacturer Code:PA546134
Catalog # PIPA546134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CTTNBP2NL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTTNBP2 N-terminal-like protein
Gene Aliases: AA552995; AA589392; AU018624; BC003236; CTTNBP2NL; KIAA1433; mKIAA1433; RGD1310822
UniProt ID: (Human) Q9P2B4, (Mouse) Q99LJ0
Entrez Gene ID: (Rabbit) 100350094, (Dog) 483222, (Human) 55917, (Horse) 100059014, (Bovine) 538981, (Rat) 310760, (Mouse) 80281, (Guinea pig) 100714721
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.