Proteintech
Manufacturer Code:255231AP150UL
Catalog # 50-173-4946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CTTNBP2NL Fusion Protein Ag22137 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTTNBP2 N terminal like CTTNBP2NL KIAA1433; CTTNBP2 N-terminal-like protein
Gene Aliases: AA552995; AA589392; AU018624; BC003236; CTTNBP2NL; KIAA1433; mKIAA1433
UniProt ID: (Human) Q9P2B4, (Mouse) Q99LJ0
Entrez Gene ID: (Human) 55917, (Mouse) 80281
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.