OriGene
Manufacturer Code:TA812843
Catalog # 50-168-4690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 114-333 of human CTSL produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Cathepsin L; Cathepsin L heavy chain; Cathepsin L light chain; Cathepsin L1; CATL CTSL1 MEP; Major excreted protein; MEP; Procathepsin L
Gene Aliases: CATL; CTSL; CTSL1; MEP
UniProt ID: (Human) P07711
Entrez Gene ID: (Human) 1514
Molecular Function:
cysteine protease
hydrolase
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.