Abnova
Manufacturer Code:H00001512M01A
Catalog # 89-021-231
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D10 |
Immunogen | CTSH (NP_004381.2 157 a.a. ~ 235 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant CTSH.
For Research Use Only
Protein Aliases: ACC-4; ACC-5; aleurain; cathepsin B3; cathepsin BA; Cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; DKFZp686B24257; MGC1519; minichain; N-benzoylarginine-beta-naphthylamide hydrolase; Pro-cathepsin H
Gene Aliases: ACC-4; ACC-5; ACC4; ACC5; CPSB; CTSH
UniProt ID: (Human) P09668
Entrez Gene ID: (Human) 1512
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.