Invitrogen
Manufacturer Code:PA520145
Catalog # PIPA520145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide from near the center of human CTRP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Complement C1q tumor necrosis factor-related protein 1; G protein coupled receptor interacting protein; G protein-coupled receptor-interacting protein; GIP; Putative secreted protein ZSIG37; UNQ310/PRO353 CTRP1 GIP ZSIG37 G protein coupled receptor interacting protein complement C1q tumor necrosis factor-related protein 1 g protein-coupled receptor-interacting protein
Gene Aliases: 1600017K21Rik; C1QTNF1; CTRP1; GIP; UNQ310/PRO353; ZSIG37
UniProt ID: (Human) Q9BXJ1, (Mouse) Q9QXP7
Entrez Gene ID: (Human) 114897, (Mouse) 56745
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.