Invitrogen
Manufacturer Code:PA530323
Catalog # PIPA530323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 199 of Human CTR9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ctr9, Paf1/RNA polymerase II complex component, homolog; RNA polymerase-associated protein CTR9 homolog; SH2 domain binding protein 1 (tetratricopeptide repeat containing); SH2 domain-binding protein 1; SH2BP1 TSBP p150 p150TSP RNA polymerase-associated protein CTR9 homolog SH2 domain binding protein 1 (tetratricopeptide repeat containing) SH2 domain-binding protein 1 TPR-containing SH2-binding phosphoprotein; TPR-containing, SH2-binding phosphoprotein
Gene Aliases: CTR9; KIAA0155; p150; p150TSP; SH2BP1; TSBP
UniProt ID: (Human) Q6PD62
Entrez Gene ID: (Human) 9646
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.