Novus Biologicals
Manufacturer Code:NBP216031
Catalog # NBP216031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human CTR9. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ctr9 Paf1/RNA polymerase II complex component homolog (S. cerevisiae) KIAA0155SH2 domain-binding protein 1 p150TSP RNA polymerase-associated protein CTR9 homolog SH2 domain binding protein 1 (tetratricopeptide repeat containing) SH2BP1p150 TPR-containing SH2-binding phosphoprotein TSBP; Ctr9, Paf1/RNA polymerase II complex component, homolog; RNA polymerase-associated protein CTR9 homolog; SH2 domain binding protein 1 (tetratricopeptide repeat containing); SH2 domain-binding protein 1; TPR-containing, SH2-binding phosphoprotein
Gene Aliases: CTR9; KIAA0155; p150; p150TSP; SH2BP1; TSBP
UniProt ID: (Human) Q6PD62
Entrez Gene ID: (Human) 9646
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.