Proteintech
Manufacturer Code:212641AP150UL
Catalog # 50-173-3343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CTR9 Fusion Protein Ag15690 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTR9 KIAA0155 p150 p150TSP SH2 domain binding protein 1 SH2BP1 TSBP; Ctr9, Paf1/RNA polymerase II complex component, homolog; p150TSP; RNA polymerase-associated protein CTR9 homolog; SH2 domain binding protein 1 (tetratricopeptide repeat containing); SH2 domain-binding protein 1; Tetratricopeptide repeat-containing, SH2-binding phosphoprotein of 150 kDa; TPR-containing, SH2-binding phosphoprotein; TPR-containing, SH2-binding phosphoprotein of 150 kDa
Gene Aliases: AA409336; CTR9; KIAA0155; mKIAA0155; p150; p150TSP; SH2BP1; TSBP; Tsp
UniProt ID: (Human) Q6PD62, (Mouse) Q62018
Entrez Gene ID: (Human) 9646, (Rat) 293184, (Mouse) 22083
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.