Novus Biologicals
Manufacturer Code:NBP105199
Catalog # NBP105199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide made to an internal portion of human CTR2 (within residues 50-100). [Swiss-Prot# O15432] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Copper transporter 2; Copper transporter 2 COPT2SLC13A2 CTR2Solute carrier family 31 member 2 hCTR2probable low affinity copper uptake protein 2 solute carrier family 31 (copper transporters) member 2; hCTR2; Probable low affinity copper uptake protein 2; solute carrier family 31 (copper transporter), member 2; solute carrier family 31 (copper transporters), member 2; Solute carrier family 31 member 2
Gene Aliases: COPT2; CTR2; hCTR2; SLC31A2
UniProt ID: (Human) O15432
Entrez Gene ID: (Human) 1318
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.