Invitrogen
Manufacturer Code:PA533031
Catalog # PIPA533031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from internal region of human CTR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: copper transport 1 homolog; Copper transporter 1; COPT1 CTR1 copper transport 1 homolog copper transporter 1 high affinity copper uptake protein 1 solute carrier family 31 member 1; hCTR1; High affinity copper uptake protein 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporters), member 1; Solute carrier family 31 member 1
Gene Aliases: COPT1; CTR1; SLC31A1
UniProt ID: (Human) O15431
Entrez Gene ID: (Human) 1317
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.