Invitrogen
Manufacturer Code:PA5115912
Catalog # PIPA5115912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC31A1 / CTR1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4930445G01Rik AI787263 AU016967 copper transport 1 homolog copper transporter 1 Copper uptake transporter 1 Copt1 CTR1 ctr-1 hCTR1 high affinity copper uptake protein high affinity copper uptake protein 1 high-affinity copper uptake protein Liver regeneration-related protein LRRGT00200 LRRGT00200 rCTR1 Slc31a1 slc31a1.L solute carrier family 13 (sodium/sulphate symporters) member 1 solute carrier family 31 (copper transporter) member 1 solute carrier family 31 (copper transporter) member 1 L homeolog solute carrier family 31 (copper transporters) member 1 Solute carrier family 31 member 1 solute carrier family 31 member 1 Xctr1 XELAEV_18038194mg zgc:76839; copper transport 1 homolog; Copper transporter 1; Copper uptake transporter 1; CTR1; hCTR1; High affinity copper uptake protein 1; Liver regeneration-related protein LRRGT00200; rCTR1; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporters), member 1; Solute carrier family 31 member 1
Gene Aliases: 4930445G01Rik; AI787263; AU016967; COPT1; CTR1; LRRGT00200; SLC31A1
UniProt ID: (Human) O15431, (Rat) Q9JK41, (Mouse) Q8K211
Entrez Gene ID: (Human) 1317, (Rat) 171135, (Mouse) 20529
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.