Invitrogen
Manufacturer Code:PA116586
Catalog # APPA6586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide of C-terminal region of human CTR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: copper transport 1 homolog; Copper transporter 1; copper transporter 1 COPT1 CTR1 hCTR1 solute carrier family 31 (copper tansporters) member 1; Copper uptake transporter 1; CTR1; hCTR1; high affinity copper uptake protein; High affinity copper uptake protein 1; high-affinity copper uptake protein; Liver regeneration-related protein LRRGT00200; rCTR1; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporters), member 1; Solute carrier family 31 member 1
Gene Aliases: 4930445G01Rik; AI787263; AU016967; COPT1; ctr-1; CTR1; LRRGT00200; SLC31A1; Xctr1; XELAEV_18038194mg; zgc:76839
UniProt ID: (Human) O15431, (Pig) Q8WNR0, (Rat) Q9JK41, (Mouse) Q8K211
Entrez Gene ID: (Human) 1317, (Pig) 397186, (Rat) 171135, (Mouse) 20529, (Zebrafish) 403028, (Xenopus) 379422
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.