Proteintech
Manufacturer Code:130851AP150UL
Catalog # 50-172-8477
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CTNS Fusion Protein Ag3904 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTNS CTNS LSB Cystinosin cystinosis nephropathic PQLC4; Cystinosin; cystinosis nephropathic; cystinosis, nephropathic
Gene Aliases: AI195360; AW049661; CTNS; CTNS-LSB; PQLC4
UniProt ID: (Human) O60931, (Mouse) P57757
Entrez Gene ID: (Human) 1497, (Mouse) 83429, (Rat) 287478
Molecular Function:
amino acid transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.