Abnova
Manufacturer Code:PAB11722
Catalog # 89-115-328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human CTNND1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyclonal antibody raised against synthetic peptide of CTNND1.
For Research Use Only
Protein Aliases: Cadherin-associated Src substrate; CAS; catenin (cadherin-associated protein), delta 1; Catenin delta-1; CTNND; KIAA0384; p120; p120 catenin; p120(cas); p120(ctn); P120CAS; P120CTN
Gene Aliases: CAS; CTNND; CTNND1; KIAA0384; p120; p120(CAS); p120(CTN); P120CAS; P120CTN
UniProt ID: (Human) O60716
Entrez Gene ID: (Human) 1500
Molecular Function:
cytoskeletal protein
intermediate filament
intermediate filament binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.