Novus Biologicals
Manufacturer Code:NBP237252
Catalog # NBP237252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E4F5 |
Immunogen | Purified recombinant fragment of human CTNNBL1 (AA: 390-557) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta-catenin-like protein 1; beta-catenin-like protein 1 C20orf33 catenin beta like 1 dJ633O20.1 FLJ21108 NAPchromosome 20 open reading frame 33 nuclear associated protein Nuclear-associated protein NYD-SP19 P14 P14L PP8304 Testis development protein NYD-SP19; catenin, beta like 1; NAP; nuclear associated protein; Nuclear-associated protein; Testis development protein NYD-SP19
Gene Aliases: C20orf33; CTNNBL1; dJ633O20.1; NAP; P14L; PP8304
UniProt ID: (Human) Q8WYA6
Entrez Gene ID: (Human) 56259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.