Novus Biologicals
Manufacturer Code:NBP142679
Catalog # NBP142679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 1 and 50 of human catenin beta like 1 using the numbering given in entry NP_110517.2 (GeneID 56259). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-catenin-like protein 1; beta-catenin-like protein 1 C20orf33 catenin beta like 1 dJ633O20.1 FLJ21108 NAPchromosome 20 open reading frame 33 nuclear associated protein Nuclear-associated protein NYD-SP19 P14 P14L PP8304 Testis development protein NYD-SP19; catenin, beta like 1; NAP; nuclear associated protein; Nuclear-associated protein; Testis development protein NYD-SP19
Gene Aliases: C20orf33; CTNNBL1; dJ633O20.1; NAP; P14L; PP8304
UniProt ID: (Human) Q8WYA6
Entrez Gene ID: (Human) 56259
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.