Proteintech
Manufacturer Code:136651AP150UL
Catalog # 50-172-8921
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CTNNBL1 Fusion Protein Ag4578 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta catenin like protein 1 C20orf33 catenin beta like 1 CTNNBL1 NAP Nuclear associated protein NYD SP19 P14L PP8304; beta catenin-like 1; Beta-catenin-like protein 1; catenin, beta like 1; NAP; nuclear associated protein; nuclear protein NAP; Nuclear-associated protein; Testis development protein NYD-SP19
Gene Aliases: 5730471K09Rik; C20orf33; CTNNBL1; dJ633O20.1; NAP; NYD-SP19; P14L; PP8304
UniProt ID: (Human) Q8WYA6, (Mouse) Q9CWL8, (Rat) Q4V8K2
Entrez Gene ID: (Human) 56259, (Mouse) 66642, (Rat) 296320
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.