Invitrogen
Manufacturer Code:PA579090
Catalog # PIPA579090
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse CTLA4 recombit protein (Position: E36-D161). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD152; CD152 antigen; CTLA-4; Cytotoxic T-lymphocyte protein 4; Cytotoxic T-lymphocyte protein 4 Cytotoxic T-lymphocyte-associated antigen 4 CTLA-4 CD152 Ctla4 Cd152; Cytotoxic T-lymphocyte-associated antigen 4; soluble form; transmembrane form
Gene Aliases: Cd152; CTLA-4; Ctla4; Ly-56; sCTLA4
UniProt ID: (Mouse) P09793
Entrez Gene ID: (Mouse) 12477, (Rat) 63835
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.