Invitrogen
Manufacturer Code:MA517587
Catalog # PIMA517587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | WKH 203 |
Immunogen | Purified rCTLA-4hlg fusion protein |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD152 antigen; CELIAC3 CTLA-4 GRD4 GSE ICOS IDDM12 CD152 isoform celiac disease 3 cytotoxic T lymphocyte associated antigen 4 short spliced form cytotoxic T-lymphocyte antigen 4; cytotoxic T-lymphocyte protein 4; soluble form; transmembrane form
Gene Aliases: Cd152; CTLA-4; sCTLA4
Entrez Gene ID: (Rat) 63835
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.