R&D Systems
Manufacturer Code:MAB386SP
Catalog # MAB386SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
In vitro Assay (IV) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1003705 |
Immunogen | Chinese Hamster Ovary cell line CHO-derived human CTLA-4 Ala37-Phe162 Accession # P16410 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: CD CD152 CD152 antigen CD152IDDM12 CD28 celiac disease 3 CELIAC3 CTLA4 CTLA-4 cytotoxic T-lymphocyte antigen 4 cytotoxic T-lymphocyte protein 4 Cytotoxic T-lymphocyte-associated antigen 4 cytotoxic T-lymphocyte-associated protein 4 cytotoxic T-lymphocyte-associated serine esterase-4 GRD4 GSE ICOS ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; CD152; CD152 isoform; celiac disease 3; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; Cytotoxic T-lymphocyte protein 4; Cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Gene Aliases: ALPS5; CD; CD152; CELIAC3; CTLA-4; CTLA4; GRD4; GSE; IDDM12
UniProt ID: (Human) P16410
Entrez Gene ID: (Human) 1493
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.