Novus Biologicals
Manufacturer Code:NBP289947H
Catalog # NB110297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 166 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human CTHRC1 (NP_612464.1 Met1-Lys243). |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: collagen triple helix repeat containing 1 collagen triple helix repeat-containing protein 1 Protein NMTC1; Collagen triple helix repeat-containing protein 1
Gene Aliases: CTHRC1; UNQ762/PRO1550
UniProt ID: (Human) Q96CG8
Entrez Gene ID: (Human) 115908
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.