Invitrogen
Manufacturer Code:PA538054
Catalog # PIPA538054
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 19 amino acid peptide near the amino terminus of human CTHRC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Collagen triple helix repeat-containing protein 1
Gene Aliases: 1110014B07Rik; CTHRC1; UNQ762/PRO1550
UniProt ID: (Human) Q96CG8, (Mouse) Q9D1D6, (Rat) Q8CG08
Entrez Gene ID: (Human) 115908, (Mouse) 68588, (Rat) 282836
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.