Invitrogen
Manufacturer Code:MA525424
Catalog # PIMA525424
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D6 |
Immunogen | Full length human recombit protein of CTH produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cystathionase; cystathionase (cystathionine gamma-lyase); Cystathionine gamma-lyase; cystathionine gamma-lyase cysteine desulfhydrase gamma-cystathionase homoserine deaminase homoserine dehydratase; cysteine desulfhydrase; Cysteine-protein sulfhydrase; Gamma-cystathionase; homoserine deaminase; homoserine dehydratase; synthase
Gene Aliases: CTH; QccE-22305
UniProt ID: (Human) P32929, (Cynomolgus monkey) Q60HG7
Entrez Gene ID: (Human) 1491, (Dog) 479991, (Cynomolgus monkey) 102117863
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.