Invitrogen
Manufacturer Code:MA538251
Catalog # PIMA538251
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT1657 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human CTH. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 0610010I13Rik AI314617 BC019483 CSE Cth CTL target antigen Cys3 cystathionase (cystathionine gamma-lyase) cystathionine gamma-lyase cysteine desulfhydrase Cysteine-protein sulfhydrase EGK_00877 Gamma-cystathionase H homoserine deaminase homoserine dehydratase MGC9471 PRB-RA Probasin-related antigen; Cystathionine gamma-lyase; Cysteine-protein sulfhydrase; Gamma-cystathionase
Gene Aliases: 0610010I13Rik; AI314617; BC019483; CSE; Cth; Cys3
UniProt ID: (Mouse) Q8VCN5
Entrez Gene ID: (Mouse) 107869
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.