Invitrogen
Manufacturer Code:MA535642
Catalog # PIMA535642
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1200 |
Immunogen | A synthesized peptide derived from human Cystathionase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 0610010I13Rik AI314617 BC019483 CSE CTH Cys3 Cystathionine gamma-lyase Cysteine-protein sulfhydrase fb58d08 Gamma-cystathionase H MGC9471 PRB-RA Probasin-related antigen wu:fb48e03 wu:fb58d08 zgc:66001 zgc:85785; CTL target antigen; cystathionase (cystathionine gamma-lyase); Cystathionine gamma-lyase; Cysteine-protein sulfhydrase; Gamma-cystathionase; PRB-RA; Probasin-related antigen
Gene Aliases: 0610010I13Rik; AI314617; BC019483; CSE; Cth; Cys3; H
UniProt ID: (Rat) P18757, (Mouse) Q8VCN5
Entrez Gene ID: (Rat) 24962, (Mouse) 107869
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.