Invitrogen
Manufacturer Code:PA542810
Catalog # PIPA542810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CTH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTL target antigen; cystathionase (cystathionine gamma-lyase); Cystathionase (cystathionine gamma-lyase) MGC9471; Cystathionine gamma-lyase; Cystathionine gamma-lyase-like protein; cysteine desulfhydrase; Cysteine-protein sulfhydrase; Gamma-cystathionase; homoserine deaminase; homoserine dehydratase; PRB-RA; Probasin-related antigen
Gene Aliases: 0610010I13Rik; AI314617; BC019483; CSE; CTH; Cys3; fb58d08; H; wu:fb48e03; wu:fb58d08; zgc:66001; zgc:85785
UniProt ID: (Bovine) Q58DW2, (Human) P32929, (Pig) Q19QT7, (Rat) P18757, (Mouse) Q8VCN5
Entrez Gene ID: (Bovine) 539159, (Human) 1491, (Dog) 479991, (Pig) 733654, (Horse) 100053614, (Rabbit) 100348407, (Rat) 24962, (Mouse) 107869, (Guinea pig) 100720299, (Zebrafish) 322055
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.