Invitrogen
Manufacturer Code:PA579088
Catalog # PIPA579088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human CTGF recombit protein (Position: R58-A349). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCN family member 2; Cellular communication network factor 2; Connective tissue growth factor; Connective tissue growth factor CCN family member 2 Hypertrophic chondrocyte-specific protein 24 Insulin-like growth factor-binding protein 8 IBP-8 IGF-binding protein 8 IGFBP-8 CTGF CCN2 HCS24 IGFBP8; Connective tissue growth-related protein; Hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP-8; Insulin-like growth factor-binding protein 8
Gene Aliases: CCN2; CTGF; CTGRP; HCS24; IGFBP8; NOV2
UniProt ID: (Human) P29279, (Rat) Q9R1E9
Entrez Gene ID: (Human) 1490, (Rat) 64032
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.