Invitrogen
Manufacturer Code:PA5111012
Catalog # PIPA5111012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human CTGF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-Nov betaIG-M2 BOS_9793 CCN family member 2 Ccn2 Cellular communication network factor 2 Connective tissue growth factor Connective tissue growth-related protein Ctgf CTGRP Fisp12 Fisp-12 Hcs24 H-CTGF Hypertrophic chondrocyte-specific protein 24 IBP-8 IGFBP8 Insulin-like growth factor-binding protein 8 MGC102839 NOV2 Protein FISP-12; CCN family member 2; Cellular communication network factor 2; Connective tissue growth factor; Hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP-8; Insulin-like growth factor-binding protein 8
Gene Aliases: CCN2; CTGF; HCS24; IGFBP8; NOV2
UniProt ID: (Human) P29279
Entrez Gene ID: (Human) 1490
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.