Invitrogen
Manufacturer Code:PA116634
Catalog # APPA6634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to a C-terminal portion of the human CTGF protein sequence (between residues 299-349). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCN family member 2; CCN2 Connective Tissue Growth Factor Hcs24 Hypertrophic chondrocyte-specific gene product 24 IGFBP8 Insulin-like Growth Factor-Binding Protein 8; Cellular communication network factor 2; Connective tissue growth factor; Connective tissue growth factor precursor (CTGF) (FISP-12 protein) (Hypertrophic chondrocyte-specific protein 24); Connective tissue growth-related protein; fibroblast inducible secreated protein; fibroblast inducible secreted protein; hypertrophic chondrocyte-specific gene product 24; Hypertrophic chondrocyte-specific protein 24; IBP-8; IGF-binding protein 8; IGFBP-8; Insulin-like growth factor-binding protein 8; Protein FISP-12
Gene Aliases: betaIG-M2; CCN2; CTGF; CTGRP; Fisp-12; Fisp12; HCS24; IGFBP8; NOV2
UniProt ID: (Human) P29279, (Mouse) P29268, (Rat) Q9R1E9
Entrez Gene ID: (Human) 1490, (Mouse) 14219, (Rat) 64032
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.