Invitrogen
Manufacturer Code:PA517143
Catalog # PIPA517143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the carboxy terminus of the human CTCF protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11 zinc finger transcriptional repressor; 11 zinc finger transcriptional repressor 11-zinc finger protein CTCFL paralog OTTHUMP00000197610 transcriptional repressor CTCF; 11-zinc finger protein; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); CTCFL paralog; Transcriptional repressor CTCF
Gene Aliases: AW108038; CTCF; MRD21
UniProt ID: (Human) P49711, (Rat) Q9R1D1, (Mouse) Q61164
Entrez Gene ID: (Human) 10664, (Rat) 83726, (Mouse) 13018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.