Novus Biologicals
Manufacturer Code:NB500194
Catalog # NB500194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 700 and the C-terminus (residue 727) of human CCCTC-binding factor using the numbering given in entry NP_006556.1 (GeneID 10664) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
For Research Use Only
Protein Aliases: 11 zinc finger transcriptional repressor; 11 zinc finger transcriptional repressor 11-zinc finger protein CCCTC-binding factor CCCTC-binding factor (zinc finger protein) CTCFL paralog transcriptional repressor CTCF; 11-zinc finger protein; CCCTC-binding factor; CCCTC-binding factor (zinc finger protein); CTCFL paralog; Transcriptional repressor CTCF
Gene Aliases: CTCF; MRD21
UniProt ID: (Human) P49711
Entrez Gene ID: (Human) 10664
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.