Invitrogen
Manufacturer Code:MA524198
Catalog # PIMA524198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 486725 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Chitobiase/CTBS Thr39-Arg385 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: chitobiase, di-N-acetyl-; CTBS chitobiase di-N-acetyl- CTBdi-N-acetylchitobiase EC 3.2.1.-; Di-N-acetylchitobiase
Gene Aliases: CTB; CTBS
UniProt ID: (Human) Q01459
Entrez Gene ID: (Human) 1486
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.