Invitrogen
Manufacturer Code:PA572338
Catalog # PIPA572338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 141-172 amino acids from the Central region of human CTBS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chitobiase, di-N-acetyl-; CTBS chitobiase di-N-acetyl- CTBdi-N-acetylchitobiase EC 3.2.1.-; Di-N-acetylchitobiase
Gene Aliases: 2210401K11Rik; CTB; CTBS
UniProt ID: (Human) Q01459, (Mouse) Q8R242
Entrez Gene ID: (Human) 1486, (Mouse) 74245
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.