Abnova
Manufacturer Code:H00001487M01
Catalog # 89-021-214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G7 |
Immunogen | CTBP1 (AAH53320 1 a.a. ~ 429 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant CTBP1.
For Research Use Only
Protein Aliases: BARS; brefeldin A-ribosylated substrate; C-terminal-binding protein 1; CtBP1; MGC104684
Gene Aliases: BARS; CTBP; CTBP1
UniProt ID: (Human) Q13363
Entrez Gene ID: (Human) 1487
Molecular Function:
dehydrogenase
oxidoreductase
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.