Invitrogen
Manufacturer Code:PA521383
Catalog # PIPA521383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 194 and 379 of CtBP1 (Uniprot IDNo.Q13363) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BARS C-terminal-binding protein 1 brefeldin A-ribosylated substrate; brefeldin A-ribosylated substrate; C-terminal-binding protein 1; CtBP1
Gene Aliases: BARS; CTBP; CTBP1
UniProt ID: (Human) Q13363
Entrez Gene ID: (Human) 1487
Molecular Function: dehydrogenase oxidoreductase transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.