Proteintech
Manufacturer Code:552791AP150UL
Catalog # 50-173-6095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CTAGE family member 5; cTAGE family member 5 CTAGE family member 5 CTAGE5 MEA11 MEA6 MGEA MGEA11 MGEA6 Protein cTAGE 5; CTAGE family member 5 ER export factor; CTAGE family, member 5; Cutaneous T-cell lymphoma-associated antigen 5; Melanoma inhibitory activity protein 2; meningioma expressed antigen 6 (coiled-coil proline-rich); Meningioma-expressed antigen 6/11; MIA protein 2; protein cTAGE-5
Gene Aliases: CTAGE5; MEA11; MEA6; MGEA; MGEA11; MGEA6; MIA2
UniProt ID: (Human) Q96PC5
Entrez Gene ID: (Human) 4253
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.