Invitrogen
Manufacturer Code:PA519062
Catalog # PIPA519062
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CGKSLESTNVTDKD) corresponding to the internal amino acids of CSRP3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac LIM domain protein; Cardiac LIM protein; CRP3; CSRP3 cysteine and glycine-rich protein 3 (cardiac LIM protein) CLP CMD1M CRP3 LMO4 MGC14488 MGC61993 MLP LIM domain only 4 cardiac LIM domain protein cardiac LIM protein cysteine and glycine-rich protein 3 cysteine-rich protein 3 muscle LIM protein; Cysteine and glycine-rich protein 3; cysteine and glycine-rich protein 3 (cardiac LIM protein); Cysteine-rich protein 3; LIM domain only 4; LIM domain protein, cardiac; Muscle LIM protein; muscle lim protein isoform
Gene Aliases: CLP; CMD1M; CMH12; CRP3; CSRP3; LMO4; MLP; MMLP
UniProt ID: (Human) P50461, (Mouse) P50462, (Rat) P50463
Entrez Gene ID: (Human) 8048, (Mouse) 13009, (Rat) 117505
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.