Novus Biologicals
Manufacturer Code:NBP106983
Catalog # NBP106983
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CGKSLESTNVTDKD corresponding to internal region according to NP_003467.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac LIM domain protein; Cardiac LIM protein; Cardiac LIM protein CLPMGC14488 CMD1Mcardiac LIM domain protein CMH12 CRP3cysteine and glycine-rich protein 3 cysteine and glycine-rich protein 3 (cardiac LIM protein) Cysteine-rich protein 3 LIM domain only 4 LIM domain protein cardiac LMO4 MLPMGC61993 Muscle LIM protein; CRP3; Cysteine and glycine-rich protein 3; cysteine and glycine-rich protein 3 (cardiac LIM protein); Cysteine-rich protein 3; LIM domain only 4; LIM domain protein, cardiac; Muscle LIM protein; muscle lim protein isoform
Gene Aliases: CLP; CMD1M; CMH12; CRP3; CSRP3; LMO4; MLP
UniProt ID: (Human) P50461
Entrez Gene ID: (Human) 8048
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.