Invitrogen
Manufacturer Code:PA577207
Catalog # PIPA577207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human CSRP2BP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADA2A-containing complex subunit 2; ATAC component 2 homolog; ATAC2; CRP2 binding partner; CRP2 binding protein; CRP2-binding partner; CRP2BP; CSRP2 binding protein; CSRP2-binding protein; cysteine and glycine-rich protein 2 binding protein; cysteine-rich protein 2 binding protein; Cysteine-rich protein 2-binding protein; Cysteine-rich protein 2-binding protein CSRP2-binding protein ADA2A-containing complex subunit 2 ATAC2 CRP2-binding partner CRP2BP CSRP2BP; Lysine acetyltransferase 14
Gene Aliases: 2510008M08Rik; ATAC2; AU023459; CRP2BP; CSRP2BP; D2Ertd473e; D2Wsu131e; dJ717M23.1; E430020F17; KAT14; PRO1194
UniProt ID: (Mouse) Q8CID0, (Human) Q9H8E8
Entrez Gene ID: (Rat) 362224, (Mouse) 228714, (Human) 57325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.