Abnova
Manufacturer Code:H00057325K
Catalog # 89-207-874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human CSRP2BP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human CSRP2BP peptide using ARM Technology.
For Research Use Only
Protein Aliases: ADA2A-containing complex subunit 2; ATAC component 2 homolog; ATAC2; CRP2 binding partner; CRP2 binding protein; CRP2-binding partner; CRP2BP; CRP2BP MGC15388 PRO1194 dJ717M23.1; CSRP2 binding protein; CSRP2-binding protein; Cysteine-rich protein 2-binding protein; Lysine acetyltransferase 14
Gene Aliases: ATAC2; CRP2BP; CSRP2BP; dJ717M23.1; KAT14; PRO1194
UniProt ID: (Human) Q9H8E8
Entrez Gene ID: (Human) 57325
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.