Proteintech
Manufacturer Code:550271AP150UL
Catalog # 50-553-888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AN2 proteoglycan; AN2/NG2 proteoglycan; Chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); Chondroitin sulfate proteoglycan NG2; CSPG4 HMW MAA MCSP MCSPG MEL CSPG MSK16 NG2; HSN tumor-specific antigen; Melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; membrane-spanning proteoglycan NG2; nerve/glial antigen 2; Proteoglycan AN2
Gene Aliases: 4732461B14Rik; AN2; CSPG4; HMW-MAA; Kiaa4232; MCSP; MCSPG; MEL-CSPG; MSK16; NG2
UniProt ID: (Human) Q6UVK1, (Rat) Q00657, (Mouse) Q8VHY0
Entrez Gene ID: (Human) 1464, (Rat) 81651, (Mouse) 121021
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.